missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)butanoic acid, 97%, Thermo Scientific™

Product Code. 10747664
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10747664 1 g 1g
10513983 250 mg 250mg
10651036 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10747664 Supplier Thermo Scientific Maybridge Supplier No. MO01273DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 35656-89-4
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD00463170
InChI Key AGIIMNQWNPUJPT-UHFFFAOYSA-N
Synonym 4-4-bromophenyl butanoic acid, 4-4-bromophenyl butyric acid, benzenebutanoic acid, 4-bromo, 4-bromo-benzenebutanoic acid, pubchem16174, acmc-209iig, 4-bromobenzenebutanoic acid, benzenebutanoic acid,4-bromo, 4-4-bromo-phenyl-butyric acid
PubChem CID 252732
IUPAC Name 4-(4-bromophenyl)butanoic acid
SMILES C1=CC(=CC=C1CCCC(=O)O)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 243.1
Percent Purity 97%
Chemical Name or Material 4-(4-Bromophenyl)butanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.