missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Chlorophenoxy)-3-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 15458315
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15458315 5 g 5g
15448315 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15458315

Brand: Thermo Scientific Alfa Aesar H33106.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57688-17-2
Molecular Formula C13H9ClF3NO
Molecular Weight (g/mol) 287.666
MDL Number MFCD01908501
InChI Key LEIVFIRDFAJXDA-UHFFFAOYSA-N
Synonym 4-4-chlorophenoxy-3-trifluoromethyl aniline, 4-4-chlorophenoxy-3-trifluoromethyl, pubchem2956, 4-4-chloranylphenoxy-3-trifluoromethyl aniline, 4-4-chlorophenoxy-3-trifluoromethyl phenylamine
PubChem CID 726092
IUPAC Name 4-(4-chlorophenoxy)-3-(trifluoromethyl)aniline
SMILES C1=CC(=CC=C1OC2=C(C=C(C=C2)N)C(F)(F)F)Cl

Specifications

Refractive Index 1.565
Quantity 5 g
UN Number UN2810
Formula Weight 287.67
Percent Purity 97%
Chemical Name or Material 4-(4-Chlorophenoxy)-3-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.