missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminodicyclohexylmethane, mixture of stereoisomers, 98+%

Product Code. 11332157
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332157 25 g 25g
11342157 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332157 Supplier Thermo Scientific Alfa Aesar Supplier No. L09598.14
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1761-71-3 | C13H26N2 | 210.365 g/mol

4,4'-Diaminodicyclohexylmethane is used as epoxy curing agents. It is a versatile intermediate to produce leather, rubber products, plastics, dyes, and photo sensitive polymers. It is used in manufacturing diisocyanates and polyamides. It is also used as lubricant additive as and corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Diaminodicyclohexylmethane is used as epoxy curing agents. It is a versatile intermediate to produce leather, rubber products, plastics, dyes, and photo sensitive polymers. It is used in manufacturing diisocyanates and polyamides. It is also used as lubricant additive as and corrosion inhibitor.

Solubility
Soluble in water. (5 g/L) at 20°C.

Notes
Incompatible materials are acids, acid chlorides, acid anhydrides, oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1761-71-3
Molecular Formula C13H26N2
Molecular Weight (g/mol) 210.365
MDL Number MFCD00001496
InChI Key DZIHTWJGPDVSGE-UHFFFAOYSA-N
Synonym 4,4'-methylenedicyclohexanamine, 4,4'-diaminodicyclohexylmethane, 4,4'-methylenebis cyclohexylamine, cyclohexanamine, 4,4'-methylenebis, wandamin hm, bis 4-aminocyclohexyl methane, bis p-aminocyclohexyl methane, 4,4'-diaminodicyclohexyl methane, pacm 20, cyclohexylamine, 4,4'-methylenebis
PubChem CID 15660
IUPAC Name 4-[(4-aminocyclohexyl)methyl]cyclohexan-1-amine
SMILES C1CC(CCC1CC2CCC(CC2)N)N

Specifications

Melting Point ∼43°C
pH 11.5
Boiling Point 325°C to 327°C
Flash Point 159°C (318°F)
Odor Amine-like
Quantity 25 g
UN Number UN2928
Beilstein 2079179
Solubility Information Soluble in water. (5g/L) at 20°C.
Formula Weight 210.37
Percent Purity ≥98%
Chemical Name or Material 4,4'-Diaminodicyclohexylmethane, mixture of stereoisomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.