missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%

Product Code. 10731632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10731632 100 g Glass bottle 100g
10586972 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10731632 Supplier Thermo Scientific Acros Supplier No. 406411000
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 81-11-8 | C14H14N2O6S2 | 370.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-11-8
Molecular Formula C14H14N2O6S2
Molecular Weight (g/mol) 370.4
MDL Number MFCD00024946
InChI Key REJHVSOVQBJEBF-OWOJBTEDSA-N
Synonym amsonic acid, flavonic acid, tinopal bhs, amsonsaeure, dasd, 4,4'-diaminostilbene-2,2'-disulfonic acid, unii-h68088wvjl, 2,2'-disulfo-4,4'-stilbenediamine, diaminostilbenedisulfonic acid, 4,4'-diaminostilbene-2,2'-disulphonic acid
PubChem CID 5284378
IUPAC Name 5-amino-2-[(E)-2-(4-amino-2-sulfophenyl)ethenyl]benzenesulfonic acid
SMILES C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)N)S(=O)(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >300°C
Color Brown-Yellow to Cream
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Beilstein 14,I,744
Merck Index 15, 590
Solubility Information Solubility in water: very slightly soluble. Other solubilities: soluble in ethanol,diethyl ether
Formula Weight 370.4
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Diaminostilbene-2, 2'-disulfonic acid, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Diaminostilbene-2,2'-disulfonic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.