missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobiphenyl, 98+%

Product Code. 11478106
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11478106

Brand: Thermo Scientific Alfa Aesar A14139.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-86-4 | C12H8Br2 | 312.00 g/mol

4,4'-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobiphenyl is commonly used as fire retardants and plasticizers. It is also have application in gas chromatography and liquid chromatography analysis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 92-86-4
Molecular Formula C12H8Br2
Molecular Weight (g/mol) 312.00
MDL Number MFCD00000101
InChI Key HQJQYILBCQPYBI-UHFFFAOYSA-N
Synonym 4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
PubChem CID 7110
IUPAC Name 1-bromo-4-(4-bromophenyl)benzene
SMILES BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifikationer

Melting Point 164°C to 169°C
Boiling Point 355°C to 360°C
Quantity 250 g
UN Number UN3152
Beilstein 2044701
Solubility Information Insoluble in water.
Formula Weight 312.02
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dibromobiphenyl

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.