missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dibromobiphenyl, 99%

Product Code. 10791165 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10791165 25 g Plastic bottle 25g
10711681 100 g Plastic bottle 100g
10551461 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10791165 Supplier Thermo Scientific Acros Supplier No. 212080250
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-86-4 | C12H8Br2 | 312.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-86-4
Molecular Formula C12H8Br2
Molecular Weight (g/mol) 312.00
MDL Number MFCD00000101
InChI Key HQJQYILBCQPYBI-UHFFFAOYSA-N
Synonym 4,4'-dibromobiphenyl, 1,1'-biphenyl, 4,4'-dibromo, p,p'-dibromobiphenyl, 4,4'-dibromodiphenyl, 1-bromo-4-4-bromophenyl benzene, 4,4'-dibromo-biphenyl, unii-pou26v2xt2, 4',4-dibromobiphenyl, biphenyl, 4,4'-dibromo, pou26v2xt2
PubChem CID 7110
IUPAC Name 1-bromo-4-(4-bromophenyl)benzene
SMILES BrC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 165°C to 169°C
Boiling Point 355°C to 360°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula BrC6H4C6H4Br
Quantity 25 g
Beilstein 05, 580
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol;,soluble in benzene
Formula Weight 312
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4, 4'-Dibromobiphenyl, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Dibromobiphenyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.