missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorobenzophenone, 99%

Product Code. 11454344
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454344 25 g 25g
11464344 100 g 100g
11474344 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454344 Supplier Thermo Scientific Alfa Aesar Supplier No. A12608.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 90-98-2 | C13H8Cl2O | 251.11 g/mol

It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Soluble in hot acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-98-2
Molecular Formula C13H8Cl2O
Molecular Weight (g/mol) 251.11
MDL Number MFCD00000623
InChI Key OKISUZLXOYGIFP-UHFFFAOYSA-N
Synonym 4,4'-dichlorobenzophenone, bis 4-chlorophenyl methanone, methanone, bis 4-chlorophenyl, bis 4-chlorophenyl ketone, dcbp, di p-chlorophenyl ketone, di 4-chlorophenyl ketone, bis p-chlorophenyl ketone, p,p'-dichlorobenzophenone, usaf do-4
PubChem CID 7034
ChEBI CHEBI:27519
IUPAC Name bis(4-chlorophenyl)methanone
SMILES ClC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 144°C to 146°C
Boiling Point 352°C to 354°C
Quantity 25 g
Beilstein 643345
Solubility Information Soluble in hot acetone.
Formula Weight 251.11
Percent Purity 99%
Chemical Name or Material 4,4'-Dichlorobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.