missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorochalcone, 98+%

Product Code. 11302966
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302966 5 g 5g
11312966 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302966 Supplier Thermo Scientific Alfa Aesar Supplier No. L10717.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19672-59-4 | C15H10Cl2O | 277.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19672-59-4
Molecular Formula C15H10Cl2O
Molecular Weight (g/mol) 277.14
MDL Number MFCD00018704
InChI Key YMEMCRBNZSLQCQ-XCVCLJGOSA-N
Synonym 4,4'-dichlorochalcone, e-1,3-bis-4-chloro-phenyl-propenone, e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl prop-2-en-1-one, 2e-1,3-bis 4-chlorophenyl-2-propen-1-one, 4',4-dichlorochalcone, e-4,4'-dichlorochalcone, trans-1,3-di-4-chlorophenyl-prop-2-en-1-one
PubChem CID 5377011
IUPAC Name (E)-1,3-bis(4-chlorophenyl)prop-2-en-1-one
SMILES ClC1=CC=C(\C=C\C(=O)C2=CC=C(Cl)C=C2)C=C1

Specifications

Melting Point 156°C to 160°C
Quantity 5 g
Beilstein 2331589
Formula Weight 277.15
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dichlorochalcone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.