missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyl sulfone, 97%

Product Code. 10401491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10401491 250 g Plastic bottle 250g
10401301 25 g Glass Bottle 25g
10145070 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10401491 Supplier Thermo Scientific Acros Supplier No. 291182500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 80-07-9 | C12H8Cl2O2S | 287.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-07-9
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.15
MDL Number MFCD00000619
InChI Key GPAPPPVRLPGFEQ-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
PubChem CID 6625
IUPAC Name 1-chloro-4-(4-chlorophenyl)sulfonylbenzene
SMILES ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 146.0°C to 151.0°C
Color White
Boiling Point 397.0°C
Flash Point 233°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 250 g
Beilstein 06, 327
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohols,slightly soluble in aqueous solvents
Formula Weight 287.16
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material 4, 4'-Dichlorodiphenyl sulfone, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.