missing translation for 'onlineSavingsMsg'
Learn More

4,4’-Dichlorodiphenyl Sulfone, TRC

Product Code. 30132992
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132992 10 g 10g
30133332 50 g 50g
30011223 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132992 Supplier Toronto Research Chemicals Supplier No. TRCD43416510G
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-07-9
Molecular Formula C12 H8 Cl2 O2 S
Molecular Weight (g/mol) 287.16
Synonym Sulfone, bis(p-chlorophenyl) (6CI,8CI), 1,1'-Sulfonylbis[4-chlorobenzene], 4,4'-Dichlorodiphenyl sulfone, 4-Chloro-1-(4-chlorophenylsulfonyl)benzene, 4-Chlorophenyl sulfone, B 0810, Bis(4-chlorophenyl) sulfone, Bis(p-chlorophenyl) sulfone, Di-p-chlorophenyl sulfone, NSC 23899, NSC 38759, NSC 50730, NSC 7207, p,p'-Dichlorodiphenyl sulfone, p-Chlorophenyl sulfone, Benzene, 1,1'-sulfonylbis[4-chloro-, 4,4'-Dichlorodiphenyl Sulfone
IUPAC Name 1-chloro-4-(4-chlorophenyl)sulfonylbenzene
SMILES Clc1ccc(cc1)S(=O)(=O)c2ccc(Cl)cc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H8Cl2O2S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8H
Formula Weight 285.9622
Percent Purity >95%
Chemical Name or Material 4,4'-Dichlorodiphenyl Sulfone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.