missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzophenone, 99%

Product Code. 10784841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10784841 100 g Glass bottle 100g
10079052 5 g Glass bottle 5g
10226150 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10784841 Supplier Thermo Scientific Acros Supplier No. 151791000
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 345-92-6 | C13H8F2O | 218.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 345-92-6
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.20
MDL Number MFCD00000353
InChI Key LSQARZALBDFYQZ-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzophenone, bis 4-fluorophenyl methanone, bis 4-fluorophenyl-methanone, methanone, bis 4-fluorophenyl, p,p'-difluorobenzophenone, di-p-fluorophenyl ketone, bis p-fluorophenyl ketone, bis 4-fluorophenyl ketone, benzophenone, 4,4'-difluoro, unii-88bnc11b9c
PubChem CID 9582
IUPAC Name bis(4-fluorophenyl)methanone
SMILES FC1=CC=C(C=C1)C(=O)C1=CC=C(F)C=C1

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point 106°C to 109°C
Color White to Yellow
Boiling Point 200°C
Loss on Drying 0.25% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (FC6H4)2CO
Quantity 100 g
Formula Weight 218.2
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4, 4'-Difluorobenzophenone, 99%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 4,4'-Difluorobenzophenone
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.