missing translation for 'onlineSavingsMsg'
Learn More

(4,4-Difluorocyclohexyl)methanol, 97%

Product Code. 15284914 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15284914 1 g 1g
15294914 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15284914 Supplier Thermo Scientific Acros Supplier No. 452470010
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 178312-48-6 | C7H12F2O | 150.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178312-48-6
Molecular Formula C7H12F2O
Molecular Weight (g/mol) 150.17
MDL Number MFCD11847775
InChI Key XJZNZSLOHZLFQP-UHFFFAOYSA-N
Synonym 4,4-difluorocyclohexyl methanol, 4,4-difluoro-cyclohexyl-methanol, 4,4-difluorocyclohexane-1-methanol, cyclohexanemethanol, 4,4-difluoro, 4,4-difluorocyclohexanemethanol, 4,4-difluoro cyclohexyl methanol, 4,4-difluoro-cyclohexyl methanol, 4,4-difluorocyclohexyl-methanol
PubChem CID 21184133
IUPAC Name (4,4-difluorocyclohexyl)methanol
SMILES OCC1CCC(F)(F)CC1

Specifications

Assay Percent Range 96% min. (GC)
Quantity 1 g
Formula Weight 150.17
Percent Purity 97%
Chemical Name or Material (4, 4-Difluorocyclohexyl)methanol
compliance-icons
Product Identifier
  • (4,4-Difluorocyclohexyl)methanol, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.