missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxybiphenyl, 99%

Product Code. 11495886
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495886 500 g 500g
11475886 25 g 25g
11485886 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495886 Supplier Thermo Scientific Alfa Aesar Supplier No. B22240.36
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-88-6 | C12H10O2 | 186.21 g/mol

4,4'-Dihydroxybiphenyl is used to prepare plastics. It serves as a laboratory reagent and an intermediate in the preparation of polyether liquid crystals. Further, it is used as an antioxidant agent for rubber and latex. In addition to this, it is used in light-colored rubber products, food packaging and medical latex products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dihydroxybiphenyl is used to prepare plastics. It serves as a laboratory reagent and an intermediate in the preparation of polyether liquid crystals. Further, it is used as an antioxidant agent for rubber and latex. In addition to this, it is used in light-colored rubber products, food packaging and medical latex products.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-88-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002348
InChI Key VCCBEIPGXKNHFW-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybiphenyl, 4,4'-biphenol, 4,4'-biphenyldiol, biphenyl-4,4'-diol, 1,1'-biphenyl-4,4'-diol, p,p'-biphenol, p,p'-diphenol, antioxidant dod, 4,4'-dioxydiphenyl, p-dihydroxydiphenyl
PubChem CID 7112
ChEBI CHEBI:34367
IUPAC Name 4-(4-hydroxyphenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)O)O

Specifications

Melting Point 279°C to 285°C
Odor Odorless
Quantity 500 g
Beilstein 1908886
Solubility Information Slightly soluble in water.
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 4,4'-Dihydroxybiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.