missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxydiphenylmethane, 98%

Product Code. 11464320
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464320 5 g 5g
11454320 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464320 Supplier Thermo Scientific Alfa Aesar Supplier No. A11417.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 620-92-8 | C13H12O2 | 200.237 g/mol

4,4'-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dihydroxydiphenylmethane, is also used as a bisphenol derivative with antioxidant activities. Bisphenol F has been reported to exhibit estrogen agonistic properties. Bisphenol F is used to make epoxy resins and coatings for various applications, such as lacquers, varnishes, liners, adhesives, plastics, water pipes, dental sealants, and food packaging.

Solubility
It is soluble in ethanol, ether, chloroform, alkali; slightly soluble in DMSO, insoluble in carbon disulfide and water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to Strong oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-92-8
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00002385
InChI Key PXKLMJQFEQBVLD-UHFFFAOYSA-N
Synonym 4,4'-methylenediphenol, bis 4-hydroxyphenyl methane, 4,4'-dihydroxydiphenylmethane, phenol, 4,4'-methylenebis, p-p-hydroxybenzyl phenol, bis p-hydroxyphenyl methane, phenol, 4,4'-methylenedi, 4,4'-methylenebisphenol, p,p'-bis hydroxyphenyl methane, 4-4-hydroxyphenyl methyl phenol
PubChem CID 12111
ChEBI CHEBI:34575
IUPAC Name 4-[(4-hydroxyphenyl)methyl]phenol
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)O)O

Specifications

Melting Point 159°C to 163°C
Quantity 5 g
Beilstein 2049425
Solubility Information It is soluble in ethanol,ether,chloroform,alkali; slightly soluble in DMSO,insoluble in carbon disulfide and water.
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material 4,4'-Dihydroxydiphenylmethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.