missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diiodobiphenyl, 99%

Product Code. 11455087
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455087 100 g 100g
11445087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455087 Supplier Thermo Scientific Alfa Aesar Supplier No. B21970.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3001-15-8 | C12H8I2 | 406.005 g/mol

4,4?-diiodobiphenyl was coupled with (trimethylsilyl) acetylene via a Sonogashira-Hagihara reaction to afford the (trimethylsilyl)ethynyl derivative. 4,4?-diiodobipheny enhanced the expression of the luciferase gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-diiodobiphenyl was coupled with (trimethylsilyl) acetylene via a Sonogashira-Hagihara reaction to afford the (trimethylsilyl)ethynyl derivative. 4,4′-diiodobipheny enhanced the expression of the luciferase gene.

Solubility
Soluble in hot Toluene(very faint turbidity).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3001-15-8
Molecular Formula C12H8I2
Molecular Weight (g/mol) 406.005
MDL Number MFCD00001057
InChI Key GPYDMVZCPRONLW-UHFFFAOYSA-N
Synonym 4,4'-diiodobiphenyl, 4,4'-diiodo-1,1'-biphenyl, 4,4'-diiododiphenyl, 1,1'-biphenyl, 4,4'-diiodo, 1-iodo-4-4-iodophenyl benzene, 4,4-diiodobiphenyl, 4-4'-iodophenyl iodobenzene, 4,4;-diiodobiphenyl, 4,4'-diiododophenyl, pract, 4-iodo-1-4-iodophenyl benzene
PubChem CID 76348
IUPAC Name 1-iodo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)I

Specifications

Melting Point 205°C to 211°C
Quantity 100 g
UN Number UN3152
Sensitivity Light sensitive
Solubility Information Soluble in hot Toluene(very faint turbidity).
Formula Weight 406
Percent Purity 99%
Chemical Name or Material 4,4'-Diiodobiphenyl
compliance-icons
Product Identifier
  • 4,4'-Diiodobiphenyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4,4'-Diiodobiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.