missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diisothiocyano-2,2'-dihydrostilbenedisulfonic Acid Disodium Salt, TRC

Product Code. 30373828
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30373828 100 mg
30387370 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30373828 Supplier Toronto Research Chemicals Supplier No. TRCD460501100MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150321-88-3
Molecular Formula C16 H10 N2 O6 S4 . 2 Na
Molecular Weight (g/mol) 500.5
Synonym 2,2'-(1,2-Ethanediyl)bis[5-isothiocyanatobenzenesulfonic Acid Disodium Salt
IUPAC Name disodium;5-isothiocyanato-2-[2-(4-isothiocyanato-2-sulfonatophenyl)ethyl]benzenesulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)c1cc(ccc1CCc2ccc(cc2S(=O)(=O)[O-])N=C=S)N=C=S

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H12N2O6S4.2Na/c19-27(20,21)15-7-13(17-9-25)5-3-11(15)1-2-12-4-6-14(18-10-26)8-16(12)28(22,23)24;;/h3-8H,1-2H2,(H,19,20,21)(H,22,23,24);;/q;2*+1/p-2
Formula Weight 499.9217
Chemical Name or Material 4,4'-Diisothiocyano-2,2'-dihydrostilbenedisulfonic Acid Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.