missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxydiphenylamine, 98%

Product Code. 11374554
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374554 5 g 5g
11364554 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11374554 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L01322.06
 Restricted Item
Estimated Shipment: 29-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 101-70-2 | C14H15NO2 | 229.28 g/mol

4,4'-Dimethoxydiphenylamine is a diphenylamine derivative used as a chemical additive for cured rubber. It is used in the synthesis of amine electron donating groups in organic donor-bridge-acceptor dyads in Dye Sensitized Solar Cells (DSSC). 4,4?-Dimethoxydiphenylamine is an electron donor and used in the synthesis of organic nonlinear optical (NLO) chromophores which exhibit high thermal and chemical stability, high degree of non-linearity and transparency. They are used in high performance photonic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxydiphenylamine is a diphenylamine derivative used as a chemical additive for cured rubber. It is used in the synthesis of amine electron donating groups in organic donor-bridge-acceptor dyads in Dye Sensitized Solar Cells (DSSC). 4,4′-Dimethoxydiphenylamine is an electron donor and used in the synthesis of organic nonlinear optical (NLO) chromophores which exhibit high thermal and chemical stability, high degree of non-linearity and transparency. They are used in high performance photonic devices.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Air Sensitive. Store under dry inert gas. Incompatible with isocyanates, halogenated organics, peroxides, phenols (acidic), epoxides, anhydrides, and acid halides.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 101-70-2
Molecular Formula C14H15NO2
Molecular Weight (g/mol) 229.28
MDL Number MFCD00014895
InChI Key VCOONNWIINSFBA-UHFFFAOYSA-N
Synonym bis 4-methoxyphenyl amine, 4,4'-dimethoxydiphenylamine, di-p-anisylamine, di-p-methoxyphenylamine, 4-methoxy-n-4-methoxyphenyl aniline, bis p-anisyl amine, bis p-methoxyphenyl amine, benzenamine, 4-methoxy-n-4-methoxyphenyl, termofleks a, bis-4-methoxy-phenyl-amine
PubChem CID 7571
IUPAC Name 4-methoxy-N-(4-methoxyphenyl)aniline
SMILES COC1=CC=C(NC2=CC=C(OC)C=C2)C=C1

Specifications

Melting Point 100°C to 104°C
Quantity 5 g
Beilstein 2214262
Sensitivity Air Sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 229.28
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethoxydiphenylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.