missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxytrityl Chloride, 98%

Product Code. 10607773 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10607773 100 g Glass bottle 100g
10407350 5 g Glass bottle 5g
10316550 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10607773 Supplier Thermo Scientific Acros Supplier No. 146021000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40615-36-9 | C21H19ClO2 | 338.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40615-36-9
Molecular Formula C21H19ClO2
Molecular Weight (g/mol) 338.83
MDL Number MFCD00008409
InChI Key JBWYRBLDOOOJEU-UHFFFAOYSA-N
Synonym 4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl
PubChem CID 96831
IUPAC Name 1-[chloro-(4-methoxyphenyl)-phenylmethyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 121°C to 127°C
Color Pink
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H5C(C6H4OCH3)2Cl
Quantity 100 g
Fieser 01,188
Formula Weight 338.82
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 4'-Dimethoxytrityl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4,4'-Dimethoxytrityl chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.