missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxytrityl Chloride, TRC

Product Code. 30134891
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134891 10 g 10g
30137559 5 g 5g
30004556 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134891 Supplier Toronto Research Chemicals Supplier No. TRCD46111510G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40615-36-9
Molecular Formula C21H19ClO2
Molecular Weight (g/mol) 338.83
Synonym 1,1'-(Chlorophenylmethylene)bis[4-methoxybenzene, 4,4'-Dimethoxytriphenylmethyl Chloride, Bis(4-methoxyphenyl)phenylmethyl Chloride, Chlorobis(4-methoxyphenyl)phenylmethane, NSC 89782, DMT-Cl, DMTCl
SMILES COc1ccc(cc1)C(Cl)(c2ccccc2)c3ccc(OC)cc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C21H19ClO2/c1-23-19-12-8-17(9-13-19)21(22,16-6-4-3-5-7-16)18-10-14-20(24-2)15-11-18/h3-15H,1-2H3
Formula Weight 338.1074
Chemical Name or Material 4,4'-Dimethoxytrityl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.