missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals

Product Code. p-7030270
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347409 5 g 5g
11357409 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347409 Supplier Thermo Scientific Alfa Aesar Supplier No. H27830.06
Add to basket
This item is not returnable. View return policy

CAS: 845-46-5 | Molecular Formula: C15H14N3NaO2 | Molecular Weight: 291.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845-46-5
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
MDL Number MFCD00020350
InChI Key OSCKRHPYZNTEIO-UHFFFAOYSA-M
Synonym p-p-dimethylaminophenylazo benzoic acid sodium salt, 4-4-dimethylaminophenylazo benzoic acid sodium salt, benzoic acid, 4-4-dimethylamino phenyl azo-, sodium salt, benzoic acid, p-p-dimethylamino phenyl azo-, sodium salt, benzoic acid, 4-2-4-dimethylamino phenyl diazenyl-, sodium salt 1:1, sodium 4-4-dimethylaminophenylazo benzoate, 4-e-4-dimethylaminophenyl azo benzoic acid
PubChem CID 23674498
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 291.28
Grade Indicator
Chemical Name or Material 4-(4-Dimethylaminophenylazo)benzoic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.