missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzhydrol, 98%

Product Code. 11307635
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11307635

Brand: Thermo Scientific Alfa Aesar L05904.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 885-77-8 | C15H16O | 212.292 g/mol

4,4'-Dimethylbenzhydrol is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethylbenzhydrol is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885-77-8
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.292
MDL Number MFCD00017216
InChI Key RGYZQSCFKFDECZ-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzhydrol, di-p-tolylmethanol, bis 4-methylphenyl methanol, benzenemethanol, 4-methyl-.alpha.-4-methylphenyl, benzhydrol, 4,4'-dimethyl, di p-tolyl methanol, 4,4-dimethylbenzhydrol, 4,4'-ditolyl carbinol, benzhydrol,4'-dimethyl
PubChem CID 279356
IUPAC Name bis(4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)O

Specifications

Melting Point 69°C to 73°C
Quantity 5 g
Beilstein 1874128
Solubility Information Insoluble in water.
Formula Weight 212.29
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethylbenzhydrol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.