missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzophenone, 98+%

Product Code. 11488066
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488066 100 g 100g
11468066 5 g 5g
11478066 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488066 Supplier Thermo Scientific Alfa Aesar Supplier No. A14111.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-97-2 | C15H14O | 210.276 g/mol

4,4'-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethylbenzophenone is used as catalytic agent and petrochemical additive. It is also used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-97-2
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00017214
InChI Key ZWPWLKXZYNXATK-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzophenone, p-tolyl ketone, bis 4-methylphenyl methanone, di-p-tolyl ketone, methanone, bis 4-methylphenyl, p,p'-dimethylbenzophenone, p,p'-dimethyl di-phenyl ketone, bis-p-tolylmethanone, benzophenone, 4,4'-dimethyl, 4,4-dimethylbenzophenone
PubChem CID 69148
IUPAC Name bis(4-methylphenyl)methanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 92°C to 96°C
Boiling Point 200°C (17 mmHg)
Quantity 100 g
Beilstein 1240527
Solubility Information Insoluble in water.
Formula Weight 210.28
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dimethylbenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.