missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzophenone, 98+%

Product Code. 10370344 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10370344 1 g Glass bottle 1g
10239980 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10370344 Supplier Thermo Scientific Supplier No. 408220010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 611-97-2 | C15H14O | 210.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-97-2
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00017214
InChI Key ZWPWLKXZYNXATK-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzophenone, p-tolyl ketone, bis 4-methylphenyl methanone, di-p-tolyl ketone, methanone, bis 4-methylphenyl, p,p'-dimethylbenzophenone, p,p'-dimethyl di-phenyl ketone, bis-p-tolylmethanone, benzophenone, 4,4'-dimethyl, 4,4-dimethylbenzophenone
PubChem CID 69148
IUPAC Name bis(4-methylphenyl)methanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 93°C to 97°C
Color Brown
Boiling Point 200°C (17.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 210.28
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethylbenzophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.