missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenoxy)benzonitrile, 98%, Thermo Scientific™

Product Code. 15485257
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485257 25 g 25g
15495257 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485257 Supplier Thermo Scientific Alfa Aesar Supplier No. H32530.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215589-24-5
Molecular Formula C13H8FNO
Molecular Weight (g/mol) 213.211
MDL Number MFCD01815090
InChI Key NXVPHQNXBDDVCT-UHFFFAOYSA-N
Synonym 4-4-fluorophenoxy benzonitrile, 4-4-fluorophenoxy benzenecarbonitrile, acmc-1chxs, 4-cyano-4'-fluorodiphenyl ether, benzonitrile,4-4-fluorophenoxy
PubChem CID 1490284
IUPAC Name 4-(4-fluorophenoxy)benzonitrile
SMILES C1=CC(=CC=C1C#N)OC2=CC=C(C=C2)F

Specifications

Melting Point 67°C to 68°C
Quantity 25 g
Formula Weight 213.21
Percent Purity 98%
Chemical Name or Material 4-(4-Fluorophenoxy)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.