missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 98+%

Product Code. 13488678 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13488678 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13488678 Supplier Thermo Scientific Acros Supplier No. 450540100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1978-61-6 | C11H12FN·HCl | 213.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1978-61-6
Melting Point 169.0°C to 173.0°C
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C11H12FN·HCl
Quantity 10 g
Synonym 4-4-fluorophenyl-1,2,3,6-tetrahydropyridine hydrochloride, 4-p-fluorophenyl-1,2,3,6-tetrahydropyridine hydrochloride, pyridine, 1,2,3,6-tetrahydro-4-p-fluorophenyl-, hydrochloride, pyridine, 4-4-fluorophenyl-1,2,3,6-tetrahydro-, hydrochloride, 4-4-fluorophenyl-1,2,3,6-tetrahydro-pyridine hydrochloride, pyridine,2,3,6-tetrahydro-, hydrochloride, 4-p-fluorophenyl-1,3,6-tetrahydropyridine hydrochloride, 4-4-fluoro-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride, 4-4-fluoro-phenyl-1,2,3,6-tetrahydropyridine hydrochloride, 4-4-fluorophenyl-1,2,5,6-tetrahydropyridine hydrochloride
InChI Key CTTBTYWBXXWBMU-UHFFFAOYSA-N
SMILES C1CNCC=C1C2=CC=C(C=C2)F.Cl
IUPAC Name 4-(4-fluorophenyl)-1,2,3,6-tetrahydropyridine;hydrochloride
Molecular Weight (g/mol) 213.68
PubChem CID 197747
Formula Weight 213.68
Percent Purity 98+%
Chemical Name or Material 4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-(4-Fluorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.