missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorophenyl)-3-thiosemicarbazide, 98%

Product Code. 15488899
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15488899

Brand: Thermo Scientific Alfa Aesar L09482.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 330-94-9 | C7H8FN3S | 185.22 g/mol

It is an important pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 330-94-9
Melting Point ∽179°C (decomposition)
Odor Odorless
Molecular Formula C7H8FN3S
MDL Number MFCD00041294
Quantity 5 g
UN Number UN2811
Beilstein 2804705
Synonym 4-4-fluorophenyl-3-thiosemicarbazide, n-4-fluorophenyl hydrazinecarbothioamide, 1-amino-3-4-fluorophenyl thiourea, 3-amino-1-4-fluorophenyl thiourea, hydrazinecarbothioamide, n-4-fluorophenyl, 4-fluorophenyl amino hydrazinomethane-1-thione, acmc-1bn9a, 4-4-fluorophenyl thiosemicarbazide, 1-azanyl-3-4-fluorophenyl thiourea
Solubility Information Insoluble in water.
InChI Key HDQJJEUAWXLNCD-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1NC(=S)NN)F
IUPAC Name 1-amino-3-(4-fluorophenyl)thiourea
Molecular Weight (g/mol) 185.22
PubChem CID 737149
Formula Weight 185.22
Percent Purity 98%
Chemical Name or Material 4-(4-Fluorophenyl)-3-thiosemicarbazide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.