missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, 97%, Thermo Scientific™

Product Code. 15258336 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15258336 1 g 1g
15294358 5 g 5g
15204368 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15258336 Supplier Thermo Scientific Acros Supplier No. 454520010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 197304-21-5
Molecular Formula C13H16O6
Molecular Weight (g/mol) 268.27
MDL Number MFCD01317805
InChI Key SEJSVFHBVLKHLB-UHFFFAOYSA-N
PubChem CID 2758419
IUPAC Name 4-(4-formyl-3,5-dimethoxyphenoxy)butanoic acid
SMILES COC1=CC(OCCCC(O)=O)=CC(OC)=C1C=O

Specifications

Melting Point 166.0°C to 169.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Synonym 4-4-formyl-3,5-dimethoxyphenoxy butanoic acid, 4-4-formyl-3,5-dimethoxyphenoxy butyric acid, 4-3',5'-dimethoxy-4'-formyl phenoxy butyric acid, 4-3', 5'-dimethoxy-4'-formyl phenoxy butyric acid, bal linker, ambotzrl-1072, 4-4-formyl-3,5-dimethoxy-phenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formyl phenoxybutyricacid, 4-3,5-dimethoxy-4-formylphenoxy butanoic acid, 4-3',5'-dimethoxy-4'-formylphenoxy butyric acid
Formula Weight 268.27
Percent Purity 97%
Chemical Name or Material 4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid
compliance-icons
Product Identifier
  • 4-(4-Formyl-3,5-dimethoxyphenoxy)butyric acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.