missing translation for 'onlineSavingsMsg'
Learn More

4,4'-(Hexafluoroisopropylidene)diphenol, 98%

Product Code. 11428297
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428297 5 g 5g
11438297 25 g 25g
11468890 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428297 Supplier Thermo Scientific Alfa Aesar Supplier No. A18370.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1478-61-1 | C15H10F6O2 | 336.23 g/mol

4,4'-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene)diphenol is used to cross-link and cure fluoroelastomers. is also used as a monomer in the synthesis of specialty polymers. Polymers containing Bisphenol AF are useful in high temperature composites, in electronic materials and other specialty applications.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1478-61-1
Molecular Formula C15H10F6O2
Molecular Weight (g/mol) 336.23
MDL Number MFCD00000439
InChI Key ZFVMWEVVKGLCIJ-UHFFFAOYSA-N
Synonym bisphenol af, 4,4'-hexafluoroisopropylidene diphenol, 2,2-bis 4-hydroxyphenyl hexafluoropropane, hexafluorobisphenol a, biphenol af, 4,4'-perfluoropropane-2,2-diyl diphenol, hexafluoroisopropylidenebis 4-hydroxybenzene, hexafluorodiphenylolpropane, hexafluoroacetone bisphenol a, 2,2-bis 4-hydroxyphenyl perfluoropropane
PubChem CID 73864
ChEBI CHEBI:72754
IUPAC Name 4-[1,1,1,3,3,3-hexafluoro-2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 158°C to 162°C
Boiling Point 400°C
Quantity 5 g
Beilstein 1891568
Solubility Information Insoluble in water.
Formula Weight 336.24
Percent Purity 98%
Chemical Name or Material 4,4'-(Hexafluoroisopropylidene)diphenol
compliance-icons
Produktidentifikator
  • 4,4'-(Hexafluoroisopropylidene)diphenol
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Ergänzende Informationen
  • MIXTURE LIST-Contain: 4,4'-(Hexafluoroisopropylidene)diphenol

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.