missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%

Product Code. 11466227
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466227 25 g 25g
11355045 1 g 1g
11456227 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11466227 Supplier Thermo Scientific Alfa Aesar Supplier No. B25150.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 122-48-5 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-48-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00048232
InChI Key OJYLAHXKWMRDGS-UHFFFAOYSA-N
Synonym zingerone, vanillylacetone, zingiberone, 4-4-hydroxy-3-methoxyphenyl butan-2-one, zingherone, 4-4-hydroxy-3-methoxyphenyl-2-butanone, gingerone, vanillyl acetone, 0-paradol, 2-butanone, 4-4-hydroxy-3-methoxyphenyl
PubChem CID 31211
ChEBI CHEBI:68657
IUPAC Name 4-(4-hydroxy-3-methoxyphenyl)butan-2-one
SMILES CC(=O)CCC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 39°C to 42°C
Quantity 25 g
Formula Weight 194.23
Percent Purity 97%
Chemical Name or Material 4-(4-Hydroxy-3-methoxyphenyl)-2-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.