missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)-2-butanone, 98%

Product Code. 11407996
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407996 100 g 100g
11497986 25 g 25g
11364709 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407996 Supplier Thermo Scientific Alfa Aesar Supplier No. A14064.22
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5471-51-2 | C10H12O2 | 164.204 g/mol

4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Hydroxyphenyl)-2-butanone is used in perfumery, in cosmetics, and as a food additive to impart a fruity odor.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5471-51-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002394
InChI Key NJGBTKGETPDVIK-UHFFFAOYSA-N
Synonym 4-4-hydroxyphenyl-2-butanone, raspberry ketone, 4-4-hydroxyphenyl butan-2-one, frambinone, oxyphenalon, rheosmin, 4-p-hydroxyphenyl-2-butanone, 4-hydroxybenzylacetone, betuligenol, rasketone
PubChem CID 21648
ChEBI CHEBI:68656
IUPAC Name 4-(4-hydroxyphenyl)butan-2-one
SMILES CC(=O)CCC1=CC=C(C=C1)O

Specifications

Melting Point 80°C to 84°C
Quantity 100 g
Beilstein 776080
Solubility Information Insoluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4-(4-Hydroxyphenyl)-2-butanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.