missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Hydroxyphenyl)cyclohexanone, 97+%

Product Code. 15492108
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492108 25 g 25g
15472108 1 g 1g
15482108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15492108 Supplier Thermo Scientific Alfa Aesar Supplier No. H55767.14
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105640-07-1 | C12H14O2 | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105640-07-1
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00210693
InChI Key SLJYPZJZQIHNGU-UHFFFAOYSA-N
Synonym 4-4-hydroxyphenyl cyclohexanone, 4-4-hydroxyphenyl cyclohexan-1-one, 4-4-hydroxyphenyl-cyclohexanone, cyclohexanone, 4-4-hydroxyphenyl, 4-4-hydroxyphenyl-1-cyclohexanone, acmc-1cukv, 4-4-oxocyclohexyl phenol, ksc495c5l, 4-4'-hydroxyphenyl-cyclohexanone
PubChem CID 4598916
IUPAC Name 4-(4-hydroxyphenyl)cyclohexan-1-one
SMILES OC1=CC=C(C=C1)C1CCC(=O)CC1

Specifications

Melting Point 170°C to 175°C
Quantity 25 g
Beilstein 3269332
Formula Weight 190.24
Percent Purity ≥97%
Chemical Name or Material 4-(4-Hydroxyphenyl)cyclohexanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.