missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Iodobenzyl)morpholine, ≥97%, Thermo Scientific™

Product Code. 10720364
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10720364 10 g 10g
10536133 1 g 1g
10198304 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10720364 Supplier Thermo Scientific Maybridge Supplier No. CC29311EA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 299159-27-6
Molecular Formula C11H14INO
Molecular Weight (g/mol) 303.143
MDL Number MFCD04974051
InChI Key IYDFKIKPNVVXQU-UHFFFAOYSA-N
Synonym 4-4-iodobenzyl morpholine, 4-4-iodophenyl methyl morpholine, 4-4-iodo-benzyl-morpholine, 4-iodobenzyl morpholine, 4-morpholinomethyliodobenzene
PubChem CID 2795500
IUPAC Name 4-[(4-iodophenyl)methyl]morpholine
SMILES C1COCCN1CC2=CC=C(C=C2)I

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 303.14
Percent Purity 95%
Chemical Name or Material 4-(4-Iodobenzyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.