missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Isopropylidenediphenol, 97%

Product Code. 13498378 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
2kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13498378 5 kg Plastic drum 5kg
10604832 100 g Plastic bottle 100g
10042020 500 g Plastic bottle 500g
10327670 2 kg Plastic bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13498378 Supplier Thermo Scientific Acros Supplier No. 158240050
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 80-05-7 | C15H16O2 | 228.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-05-7
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00002366
InChI Key IISBACLAFKSPIT-UHFFFAOYSA-N
Synonym bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian
PubChem CID 6623
ChEBI CHEBI:33216
IUPAC Name 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 155.0°C to 158.0°C
Color Yellow to Orange
Boiling Point 220.0°C (4.0 mmHg)
Flash Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic drum
Linear Formula (CH3)2C(C6H4OH)2
Quantity 5 kg
Beilstein 06,1011
Merck Index 15,1298
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in dilute alkalies, soluble in alcohol, slightly soluble in ccl4
Formula Weight 228.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4,4′-Isopropylidenediphenol, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4'-Isopropylidenediphenol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361f-Suspected of damaging fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.