missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyl)butyric acid, 98%

Product Code. 15417098
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417098 1 g 1g
15427098 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417098 Supplier Thermo Scientific Alfa Aesar Supplier No. H60613.03
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4521-28-2 | C11H14O3 | 194.23 g/mol

4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(4-Methoxyphenyl)butyric acid on demethylation with pyridinium hydrochloride affords 4-hydroxyphenylbutyric acid, a key starting material for the synthesis of preclinical candidate LY518674.

Solubility
Slightly soluble in methanol. Insoluble in chloroform.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4521-28-2
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00004404
InChI Key LZHMNCJMXQKSBY-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl butyric acid, 4-4-methoxyphenyl butanoic acid, benzenebutanoic acid, 4-methoxy, 4-4-methoxyphenyl butyricacid, 4-4-methoxyphenyl-1-butyric acid, p-methoxy-g-phenylbutyric acid, pubchem15207, 4-methoxybenzenebutyric acid, 4-p-methoxyphenylbutyric acid
PubChem CID 78280
IUPAC Name 4-(4-methoxyphenyl)butanoic acid
SMILES COC1=CC=C(C=C1)CCCC(=O)O

Specifications

Melting Point 56°C to 59°C
Quantity 1 g
Beilstein 2416568
Solubility Information Slightly soluble in methanol. Insoluble in chloroform.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material 4-(4-Methoxyphenyl)butyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.