missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester, Thermo Scientific™

Product Code. 15479155
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479155 1 g 1g
15489155 5 g 5g
15499155 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15479155 Supplier Thermo Scientific Alfa Aesar Supplier No. H51116.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871366-38-0
Molecular Formula C20H24BNO3
Molecular Weight (g/mol) 337.226
MDL Number MFCD10567027
InChI Key FEZZNCDZVGVBHZ-UHFFFAOYSA-N
Synonym 4-4-methoxyphenylimino methyl phenylboronic acid pinacol ester, 4-4-methoxyphenyl iminomethylphenylboronic acid pinacol ester, 4-4-methoxyphenyliminomethyl benzeneboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylidene-4-methoxybenzenamine, 1e-n-4-methoxyphenyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, amtb092, 4-4-methoxyphenyl iminomethyl phenyl boronic acid pinacol ester, 1-methoxyphenyliminomethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene
PubChem CID 46739070
IUPAC Name N-(4-methoxyphenyl)-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=NC3=CC=C(C=C3)OC

Specifications

Quantity 1 g
Formula Weight 337.22
Chemical Name or Material 4-(4-Methoxyphenyliminomethyl)benzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.