missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2,6-di-tert-butylphenol), 98%

Product Code. 11453158
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453158 100 g 100g
11389439 50 g 50g
11463158 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453158 Supplier Thermo Scientific Alfa Aesar Supplier No. A18910.22
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-82-1 | C29H44O2 | 424.669 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-82-1
Molecular Formula C29H44O2
Molecular Weight (g/mol) 424.669
MDL Number MFCD00008822
InChI Key MDWVSAYEQPLWMX-UHFFFAOYSA-N
Synonym 4,4'-methylenebis 2,6-di-tert-butylphenol, bimox m, ionox 220, binox m, antioxidant e 702, mb 1 antioxidant, ethyl 702, ionox 220 antioxidant, etil 702, 4,4'-methylenebis 2,6-di-t-butylphenol
PubChem CID 8372
ChEBI CHEBI:34369
IUPAC Name 2,6-ditert-butyl-4-[(3,5-ditert-butyl-4-hydroxyphenyl)methyl]phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 154°C to 158°C
Boiling Point 289°C (40 mmHg)
Quantity 100 g
Beilstein 1916919
Formula Weight 424.67
Percent Purity 98%
Chemical Name or Material 4,4'-Methylenebis(2,6-di-tert-butylphenol)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.