missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(2-methylcyclohexylamine), 99%, mixture of isomers

Product Code. 15224814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15224814 1 L Glass bottle 1 L
15234814 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15224814 Supplier Thermo Scientific Acros Supplier No. 450240010
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6864-37-5 | C15H30N2 | 238.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6864-37-5
Molecular Formula C15H30N2
Molecular Weight (g/mol) 238.42
MDL Number MFCD00075535
InChI Key IGSBHTZEJMPDSZ-UHFFFAOYNA-N
Synonym 4,4'-methylenebis 2-methylcyclohexanamine, bis 4-amino-3-methylcyclohexyl methane, cyclohexanamine, 4,4'-methylenebis 2-methyl, 4,4'-methylenebis 2-methylcyclohexylamine, 3,3'-dimethyl-4,4'-diaminodicyclohexylmethane, 2,2'-dimethyl-4,4'-methylenebis cyclohexylamine, 4-4-amino-3-methylcyclohexyl methyl-2-methylcyclohexan-1-amine, 4,4'-methylenebis 2-methylcyclohexyl-amine, 2,2'-dimethyl-4,4'methylenebis cyclohexylamine
PubChem CID 91555
IUPAC Name 4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine
SMILES CC1CC(CC2CCC(N)C(C)C2)CCC1N

Specifications

Density 0.9400g/mL
Boiling Point 93.0°C to 100.0°C
Flash Point 113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 L
Specific Gravity 0.94
Viscosity 142 mPa.s (20°C)
Formula Weight 238.41
Percent Purity 99%
Chemical Name or Material 4, 4'-Methylenebis(2-methylcyclohexylamine)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4'-Methylenebis(2-methylcyclohexylamine)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.