missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Methylenebis(N,N-dimethylaniline), 98+%

Product Code. 11468387
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468387 50 g 50g
11478387 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11468387 Supplier Thermo Scientific Alfa Aesar Supplier No. A18466.18
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 101-61-1 | C17H22N2 | 254.377 g/mol

4,4'-Methylenebis(N,N-dimethylaniline)is used as a chemical intermediate in the manufacture of dyes and for its hydrochloric salt and is also used as an analytical reagent in the determination of lead.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Methylenebis(N,N-dimethylaniline)is used as a chemical intermediate in the manufacture of dyes and for its hydrochloric salt and is also used as an analytical reagent in the determination of lead.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-61-1
Molecular Formula C17H22N2
Molecular Weight (g/mol) 254.377
MDL Number MFCD00008317
InChI Key JNRLEMMIVRBKJE-UHFFFAOYSA-N
Synonym 4,4'-methylenebis n,n-dimethylaniline, tetra-base, tetrabase, michler's base, methane base, methylene base, michler's hydride, michler's methane, 4,4'-bis dimethylamino diphenylmethane, reduced michler's ketone
PubChem CID 7567
ChEBI CHEBI:34370
IUPAC Name 4-[[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)CC2=CC=C(C=C2)N(C)C

Specifications

Melting Point 85°C to 88°C
Boiling Point 155°C to 157°C (0.1 mmHg)
Flash Point 178°C (352°F)
Quantity 50 g
UN Number UN3077
Beilstein 479956
Merck Index 14,6176
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 254.38
Percent Purity ≥98%
Chemical Name or Material 4,4'-Methylenebis(N,N-dimethylaniline)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.