missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylperhydro-1,4-diazepin-1-yl)benzoic acid hydrochloride hydrate, 95%, Thermo Scientific™

Product Code. 10173424
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173424 250 mg Amber Glass Bottle 250mg
10482464 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173424 Supplier Thermo Scientific Maybridge Supplier No. CC60351CB
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 303134-03-4
Molecular Formula C13H19ClN2O2
Molecular Weight (g/mol) 270.757
MDL Number MFCD09064999
InChI Key KJZVHMAKWMCIBB-UHFFFAOYSA-N
Synonym 4-4-methyl-1,4-diazepan-1-yl benzoic acid hydrochloride, 4-4-methyl-1,4-diazepan-1-yl benzoic acid-hydrogen chloride 1/1
PubChem CID 24229616
IUPAC Name 4-(4-methyl-1,4-diazepan-1-yl)benzoic acid;hydrochloride
SMILES CN1CCCN(CC1)C2=CC=C(C=C2)C(=O)O.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 288.78
Percent Purity 95%
Chemical Name or Material 4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzoic acid hydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.