missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methylperhydro-1,4-diazepin-1-yl)benzoic acid hydrochloride hydrate, 95%, Thermo Scientific™

Product Code. p-3485578
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10173424

Brand: Thermo Scientific Maybridge CC60351CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 303134-03-4
Molecular Formula C13H19ClN2O2
Molecular Weight (g/mol) 270.757
MDL Number MFCD09064999
InChI Key KJZVHMAKWMCIBB-UHFFFAOYSA-N
Synonym 4-4-methyl-1,4-diazepan-1-yl benzoic acid hydrochloride, 4-4-methyl-1,4-diazepan-1-yl benzoic acid-hydrogen chloride 1/1
PubChem CID 24229616
IUPAC Name 4-(4-methyl-1,4-diazepan-1-yl)benzoic acid;hydrochloride
SMILES CN1CCCN(CC1)C2=CC=C(C=C2)C(=O)O.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 288.78
Percent Purity 95%
Chemical Name or Material 4-(4-Methylperhydro-1, 4-diazepin-1-yl)benzoic acid hydrochloride hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.