missing translation for 'onlineSavingsMsg'
Learn More

{4-[(4-Methylperhydro-1,4-diazepin-1-yl)methyl]phenyl}methanol, 97%, Thermo Scientific™

Product Code. p-3522392
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10778645 1 g Amber Glass Bottle 1g
11346795 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10778645 Supplier Thermo Scientific Maybridge Supplier No. CC55809DA
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-50-0
Molecular Formula C14H22N2O
Molecular Weight (g/mol) 234.343
MDL Number MFCD09064973
InChI Key YQUWAYPNHCWYNU-UHFFFAOYSA-N
Synonym 4-4-methylperhydro-1,4-diazepin-1-yl methyl phenyl methanol, 4-4-methyl-1,4-diazepan-1-yl methyl phenyl methanol, 4-4-methyl-1,4-diazaperhydroepinyl methyl phenyl methan-1-ol, benzenemethanol,4-hexahydro-4-methyl-1h-1,4-diazepin-1-yl methyl
PubChem CID 24229552
IUPAC Name [4-[(4-methyl-1,4-diazepan-1-yl)methyl]phenyl]methanol
SMILES CN1CCCN(CC1)CC2=CC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 234.34
Percent Purity 97%
Chemical Name or Material {4-[(4-Methylperhydro-1, 4-diazepin-1-yl)methyl]phenyl}methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.