missing translation for 'onlineSavingsMsg'
Learn More

4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochlorideHemihydrate, 95%, Thermo Scientific™

Product Code. 10451513
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10451513 250 mg Amber Glass Bottle 250mg
10330943 1 g Amber Glass Bottle 1g
10295754 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10451513 Supplier Thermo Scientific Maybridge Supplier No. CC45651CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 106261-49-8
Molecular Formula C13H20Cl2N2O2
Molecular Weight (g/mol) 307.215
InChI Key ISHROKOWRJDOSN-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl methyl benzoic acid dihydrochloride, 4-4-methyl-1-piperazinyl methyl benzoic acid dihydrochloride, 4-4-methylpiperazin-1-yl methyl, 4-4-methylpiperazinomethyl benzoic acid dihydrochloride, benzoic acid, 4-4-methyl-1-piperazinyl methyl-, dihydrochloride, zlchem 396, ksc909c2h, 4-4-methyl piperazino methyl benzoic acid 2hcl, 4-4-methylpiperazin-1-yl methyl benzoic acid dih
PubChem CID 10335346
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid;dihydrochloride
SMILES CN1CCN(CC1)CC2=CC=C(C=C2)C(=O)O.Cl.Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 316.23
Percent Purity 95%
Chemical Name or Material 4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride 0.5 hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.