missing translation for 'onlineSavingsMsg'
Learn More

4-(4-morpholinylmethyl)benzonitrile, 97%, Thermo Scientific™

Product Code. 10154252 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10154252 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10154252 Supplier Thermo Scientific Acros Supplier No. 376330010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 37812-51-4
CAS Min % 96.0
CAS Max % 100.0
Melting Point 84.0°C to 89.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C12H14N2O
MDL Number MFCD00454271
Quantity 1 g
Synonym 4-morpholinomethyl benzonitrile, 4-morpholin-4-ylmethyl benzonitrile, usaf a-3730, p-tolunitrile, alpha-4-morpholinyl, alpha-4-morpholinyl-p-tolunitrile, benzonitrile, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl benzonitrile, 4-morpholin-4-ylmethyl benzenecarbonitrile, 4-morpholin-4-ylmethyl-benzonitrile, benzonitrile,4-4-morpholinylmethyl
InChI Key BCZLYNFDOJXWGN-UHFFFAOYSA-N
SMILES C1COCCN1CC2=CC=C(C=C2)C#N
IUPAC Name 4-(morpholin-4-ylmethyl)benzonitrile
Molecular Weight (g/mol) 202.26
PubChem CID 216864
Formula Weight 202.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Morpholinylmethyl)benzonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-(4-Morpholinylmethyl)benzonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.