missing translation for 'onlineSavingsMsg'
Learn More

4-[5-(4-Dimethylaminophenyl)-2-oxazolyl]benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15469965
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469965 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15469965 Supplier Thermo Scientific Alfa Aesar Supplier No. H52632.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 380499-66-1
Molecular Formula C17H17BN2O3
MDL Number MFCD08689479
Quantity 250 mg
Synonym 4-5-4-dimethylamino phenyl oxazol-2-yl phenyl boronic acid, 4-5-4-dimethylaminophenyl oxazol-2-yl phenylboronic acid, 4-5-4-dimethylamino phenyl-1,3-oxazol-2-yl phenylboronic acid, boronic acid, 4-5-4-dimethylamino phenyl-2-oxazolyl phenyl, acmc-209ixc, 4-5-4-dimethylaminophenyl oxazol-2-yl phenylboronic acid,, 4-5-4-dimethylamino phenyl oxazol-2-yl phenyl boronicacid, 4-5-4-dimethylaminophenyl oxazol-2-yl benzeneboronic acid, 4-5-4-dimethylamino phenyl oxazole-2-yl phenylboranic acid
InChI Key VONPEMBRCXBCNJ-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C2=NC=C(O2)C3=CC=C(C=C3)N(C)C)(O)O
IUPAC Name [4-[5-[4-(dimethylamino)phenyl]-1,3-oxazol-2-yl]phenyl]boronic acid
Molecular Weight (g/mol) 308.144
PubChem CID 10335403
Formula Weight 308.14
Percent Purity 97%
Chemical Name or Material 4-[5-(4-Dimethylaminophenyl)-2-oxazolyl]benzeneboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.