missing translation for 'onlineSavingsMsg'
Learn More

4',5,7-Trihydroxyflavanone, 97%

Product Code. 11322287
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322287 10 g 10g
11312287 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322287 Supplier Thermo Scientific Alfa Aesar Supplier No. L09834.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67604-48-2 | C15H12O5 | 272.256 g/mol

(±)-Naringenin is an antioxidant flavonoid. It has anti-inflammatory and antitumor properties. (±)-Naringenin induces apoptosis. It stimulates DNA repair following oxidative damage. (±)-Naringenin inhibits the activity of PI 3-kinase (PI(3)K).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-Naringenin is an antioxidant flavonoid. It has anti-inflammatory and antitumor properties. (±)-Naringenin induces apoptosis. It stimulates DNA repair following oxidative damage. (±)-Naringenin inhibits the activity of PI 3-kinase (PI(3)K).

Solubility
Soluble in ethanol (50 mg/ml), DMSO, methanol, and ammonium hydroxide (50 mg/ml). Insoluble in water (almost)

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67604-48-2
Molecular Formula C15H12O5
Molecular Weight (g/mol) 272.256
MDL Number MFCD00006844
InChI Key FTVWIRXFELQLPI-UHFFFAOYSA-N
Synonym naringenin, 5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, naringenine, 4',5,7-trihydroxyflavanone, +/--naringenin, naringetol, salipurpol, --naringenin, narigenin, salipurol
PubChem CID 932
ChEBI CHEBI:50202
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
SMILES C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O

Specifications

Melting Point 248°C to 251°C
Quantity 10 g
Merck Index 14,6424
Solubility Information Soluble in ethanol (50mg/ml),DMSO,methanol,and ammonium hydroxide (50mg/ml). Insoluble in water (almost)
Formula Weight 272.26
Percent Purity 97%
Chemical Name or Material 4',5,7-Trihydroxyflavanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.