missing translation for 'onlineSavingsMsg'
Learn More

4,5',8-Trimethylpsoralen, 99%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63226.03

Quantity 1g

 View more versions of this product

Product Code. 15415489

  • 83.00 € / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4,5′,8-Trimethylpsoralen is used for treatment of psoriasis, and as a photochemical crosslinker of DNA used as a probe for nucleic acid structure, used to crosslink DNA onto mica surfaces, mutagenesis of N2 worms.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

3902-71-4
228.25
FMHHVULEAZTJMA-UHFFFAOYSA-N
5585
2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one
C14H12O3
MFCD00005010
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
CHEBI:28329
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1

Specifications

3902-71-4
MFCD00005010
UN1759
14,9735
Soluble in chloroform
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1
228.25
CHEBI:28329
99%
C14H12O3
1g
221723
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
FMHHVULEAZTJMA-UHFFFAOYSA-N
2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one
5585
228.25
4,5',8-Trimethylpsoralen
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.