Learn More
4,5',8-Trimethylpsoralen, 99%, Thermo Scientific™
83.00 € - 343.00 €
Chemical Identifiers
| CAS | 3902-71-4 |
|---|---|
| Molecular Formula | C14H12O3 |
| Molecular Weight (g/mol) | 228.25 |
| MDL Number | MFCD00005010 |
| InChI Key | FMHHVULEAZTJMA-UHFFFAOYSA-N |
| Synonym | trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen |
| PubChem CID | 5585 |
| ChEBI | CHEBI:28329 |
| IUPAC Name | 2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one |
| SMILES | CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15415489
|
Thermo Scientific Alfa Aesar
J63226.03 |
1g |
83.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15425489
|
Thermo Scientific Alfa Aesar
J63226.06 |
5g |
343.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4,5′,8-Trimethylpsoralen is used for treatment of psoriasis, and as a photochemical crosslinker of DNA used as a probe for nucleic acid structure, used to crosslink DNA onto mica surfaces, mutagenesis of N2 worms.
Solubility
Soluble in chloroform
Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| C14H12O3 | |
| MFCD00005010 | |
| trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen | |
| CHEBI:28329 | |
| CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1 |
Specifications
| 3902-71-4 | |
| MFCD00005010 | |
| 221723 | |
| trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen | |
| FMHHVULEAZTJMA-UHFFFAOYSA-N | |
| 2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one | |
| 5585 | |
| 228.25 | |
| 4,5',8-Trimethylpsoralen |
| C14H12O3 | |
| UN1759 | |
| 14,9735 | |
| Soluble in chloroform | |
| CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1 | |
| 228.25 | |
| CHEBI:28329 | |
| 99% |
Safety and Handling
GHS H Statement
H314-H318-H351
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.
P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H351
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
missing translation for 'einecsNumber' : 223-459-0
missing translation for 'rtecsNumber' : LV1576000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.