missing translation for 'onlineSavingsMsg'
Learn More

4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene, 97%

Product Code. 15437565
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
15437565 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 15437565

Brand: Thermo Scientific Alfa Aesar H26241.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 161265-03-8 | C39H32OP2 | 578.63 g/mol

4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.

Solubility
Soluble in organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 161265-03-8
Melting Point 224°C to 228°C
Molecular Formula C39H32OP2
MDL Number MFCD00233866
Quantity 25 g
Synonym xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9 h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene
Solubility Information Soluble in organic solvents.
InChI Key CXNIUSPIQKWYAI-UHFFFAOYSA-N
SMILES CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name (5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane
Molecular Weight (g/mol) 578.63
PubChem CID 636044
Formula Weight 578.62
Percent Purity 97%
Chemical Name or Material 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.