missing translation for 'onlineSavingsMsg'
Learn More

4,5-Desisopropylidene Topiramate, TRC

Product Code. 17961355
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17961355 10 mg 10mg
30133809 100 mg 100mg
17923385 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17961355 Supplier Toronto Research Chemicals Supplier No. TRCD29017510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106881-41-8
Formula Weight 299.0675
InChI Formula InChI=1S/C9H17NO8S/c1-8(2)17-7-6(12)5(11)3-15-9(7,18-8)4-16-19(10,13)14/h5-7,11-12H,3-4H2,1-2H3,(H2,10,13,14)/t5-,6-,7+,9+/m1/s1
IUPAC Name [(3aS,6R,7R,7aS)-6,7-dihydroxy-2,2-dimethyl-5,6,7,7a-tetrahydro-[1,3]dioxolo[4,5-b]pyran-3a-yl]methyl sulfamate
Molecular Formula C9 H17 N O8 S
Molecular Weight (g/mol) 299.3
Percent Purity >95%
SMILES CC1(C)O[C@H]2[C@H](O)[C@H](O)CO[C@@]2(COS(=O)(=O)N)O1
Synonym 2,3-O-(1-Methylethylidene)-β-D-fructopyranose 1-Sulfamate, 5H-1,3-Dioxolo[4,5-b]pyran, β-D-fructopyranose deriv.
Chemical Name or Material 4,5-Desisopropylidene Topiramate
Grade HPLC
Show More Show Less

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.