missing translation for 'onlineSavingsMsg'
Learn More

4',5-Dihydroxy-7-methoxyflavone, 97%

Product Code. 11339025
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339025 25 mg 25mg
11349025 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339025 Supplier Thermo Scientific Alfa Aesar Supplier No. L14169.MB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 437-64-9 | C16H12O5 | 284.267 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-64-9
Molecular Formula C16H12O5
Molecular Weight (g/mol) 284.267
MDL Number MFCD00017452
InChI Key JPMYFOBNRRGFNO-UHFFFAOYSA-N
Synonym genkwanin, gengkwanin, 4',5-dihydroxy-7-methoxyflavone, 5,4'-dihydroxy-7-methoxyflavone, 7-o-methylapigenin, 7-methylapigenin, apigenin 7-methyl ether, puddumetin, gonkwanin, unii-5k3i5d6b2b
PubChem CID 5281617
ChEBI CHEBI:75718
IUPAC Name 5-hydroxy-2-(4-hydroxyphenyl)-7-methoxychromen-4-one
SMILES COC1=CC(=C2C(=C1)OC(=CC2=O)C3=CC=C(C=C3)O)O

Specifications

Melting Point 288°C to 290°C
Quantity 25 mg
Beilstein 292549
Formula Weight 284.27
Percent Purity 97%
Chemical Name or Material 4',5-Dihydroxy-7-methoxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.